Assay Details
| Gene Symbol: Znf652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503944.2 | XP_003503992.1 | 3 - 4 | 1556 | 89 | |
| GenBank mRNA | JI886191.1 | - | 2 - 3 | 183 | 89 | |
| JP048343.1 | - | 2 - 3 | 183 | 89 | ||
More Information
Set Membership: |
|

Znf652
Genomic Map