Assay Details
| Gene Symbol: Sdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653636.1 | XP_007651826.1 | 6 - 7 | 818 | 63 | |
| GenBank mRNA | JI884667.1 | - | 6 - 7 | 801 | 63 | |
More Information
Set Membership: |
|

Sdhb
Genomic Map