Assay Details
| Gene Symbol: Fcrla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647416.1 | XP_007645606.1 | 5 - 6 | 813 | 73 | |
| XM_007647417.1 | XP_007645607.1 | 4 - 5 | 792 | 73 | ||
| XM_007647418.1 | XP_007645608.1 | 3 - 4 | 525 | 73 | ||
| XM_007647419.1 | XP_007645609.1 | 3 - 4 | 387 | 73 | ||
| XM_007647420.1 | XP_007645610.1 | 2 - 3 | 366 | 73 | ||
More Information
Set Membership: |
|

Fcrla
Genomic Map