Assay Details
| Gene Symbol: Lactb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646013.1 | XP_007644203.1 | 2 - 3 | 507 | 68 | |
| GenBank mRNA | JP042711.1 | - | 2 - 3 | 507 | 68 | |
More Information
Set Membership: |
|

Lactb2
Genomic Map