Assay Details
| Gene Symbol: Golga7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497785.1 | XP_003497833.1 | 3 - 4 | 381 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golga7b| Gene Symbol: Golga7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497785.1 | XP_003497833.1 | 3 - 4 | 381 | 92 | |