Assay Details
| Gene Symbol: Cdyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649400.1 | XP_007647590.1 | 2 - 3 | 671 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdyl2| Gene Symbol: Cdyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649400.1 | XP_007647590.1 | 2 - 3 | 671 | 55 | |
Set Membership: |
|