Assay Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499357.2 | XP_003499405.1 | 10 - 11 | 1172 | 97 | |
| GenBank mRNA | JI875425.1 | - | 4 - 5 | 397 | 97 | |
More Information
Set Membership: |
|

Sms
Genomic Map