Assay Details
| Gene Symbol: Cdrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649526.1 | XP_007647716.1 | 2 - 3 | 159 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdrt4| Gene Symbol: Cdrt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649526.1 | XP_007647716.1 | 2 - 3 | 159 | 62 | |
Set Membership: |
|