Assay Details
| Gene Symbol: Prelid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007623658.1 | XP_007621848.1 | 2 - 3 | 470 | 64 | |
| GenBank mRNA | JI896801.1 | - | 1 | 459 | 64 | |
More Information
Set Membership: |
|

Prelid1
Genomic Map