Assay Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500114.2 | XP_003500162.1 | 7 - 8 | 844 | 65 | |
| GenBank mRNA | JI892953.1 | - | 7 - 8 | 745 | 65 | |
| JP066025.1 | - | 7 - 8 | 745 | 65 | ||
More Information
Set Membership: |
|

Acbd6
Genomic Map