Assay Details
| Gene Symbol: Ephx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502344.2 | XP_003502392.1 | 6 - 7 | 854 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ephx4| Gene Symbol: Ephx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502344.2 | XP_003502392.1 | 6 - 7 | 854 | 76 | |
Set Membership: |
|