Assay Details
| Gene Symbol: Znf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513956.2 | XP_003514004.1 | 12 - 13 | 934 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf185| Gene Symbol: Znf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513956.2 | XP_003514004.1 | 12 - 13 | 934 | 64 | |
Set Membership: |
|