Assay Details
| Gene Symbol: Ppp1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498387.2 | XP_003498435.1 | 4 - 5 | 530 | 65 | |
| XM_007627047.1 | XP_007625237.1 | 3 - 4 | 454 | 65 | ||
| GenBank mRNA | JI881841.1 | - | 2 - 3 | 377 | 65 | |
| JP044655.1 | - | 4 - 5 | 542 | 65 | ||
More Information
Set Membership: |
|

Ppp1r8
Genomic Map