Assay Details
| Gene Symbol: LOC100763140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510937.1 | XP_003510985.1 | 10 - 11 | 1426 | 100 | |
| XM_003510938.1 | XP_003510986.1 | 9 - 10 | 1327 | 100 | ||
More Information
Set Membership: |
|

LOC100763140
Genomic Map