Assay Details
| Gene Symbol: LOC100763140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510937.1 | XP_003510985.1 | 7 - 8 | 1061 | 100 | |
| XM_003510938.1 | XP_003510986.1 | 6 - 7 | 962 | 100 | ||
More Information
Set Membership: |
|

LOC100763140
Genomic Map