Assay Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495411.2 | XP_003495459.1 | 2 - 3 | 346 | 76 | |
| GenBank mRNA | JP057189.1 | - | 2 - 3 | 335 | 76 | |
More Information
Set Membership: |
|

Umps
Genomic Map