Assay Details
| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498945.2 | XP_003498993.1 | 3 - 4 | 693 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h12d| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498945.2 | XP_003498993.1 | 3 - 4 | 693 | 92 | |
Set Membership: |
|