Assay Details
| Gene Symbol: Dhcr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505694.2 | XP_003505742.1 | 7 - 8 | 978 | 62 | |
| XM_007646172.1 | XP_007644362.1 | 8 - 9 | 1233 | 62 | ||
| XM_007646173.1 | XP_007644363.1 | 9 - 10 | 1279 | 62 | ||
| XM_007646174.1 | XP_007644364.1 | 7 - 8 | 1121 | 62 | ||
| XM_007646175.1 | XP_007644365.1 | 8 - 9 | 1253 | 62 | ||
| GenBank mRNA | JI880296.1 | - | 6 - 7 | 953 | 62 | |
| JP064895.1 | - | 6 - 7 | 954 | 62 | ||
More Information
Set Membership: |
|

Dhcr7
Genomic Map