Assay Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508476.2 | XP_003508524.1 | 5 - 6 | 735 | 67 | |
| GenBank mRNA | JI882788.1 | - | 5 - 6 | 756 | 67 | |
| JP043884.1 | - | 5 - 6 | 756 | 67 | ||
More Information
Set Membership: |
|

Clic1
Genomic Map