Assay Details
| Gene Symbol: Pvrig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651044.1 | XP_007649234.1 | 4 - 5 | 864 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pvrig| Gene Symbol: Pvrig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651044.1 | XP_007649234.1 | 4 - 5 | 864 | 82 | |
Set Membership: |
|