Assay Details
| Gene Symbol: LOC100773575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514848.2 | XP_003514896.1 | 2 - 3 | 528 | 81 | |
| XM_007654895.1 | XP_007653085.1 | 2 - 3 | 633 | 81 | ||
| GenBank mRNA | JI863150.1 | - | 2 - 3 | 502 | 81 | |
| JP041729.1 | - | 2 - 3 | 501 | 81 | ||
More Information
Set Membership: |
|

LOC100773575
Genomic Map