Assay Details
| Gene Symbol: Lpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499928.2 | XP_003499976.1 | 2 - 3 | 486 | 62 | |
| GenBank mRNA | JI882098.1 | - | 2 - 3 | 254 | 62 | |
| JP050510.1 | - | 2 - 3 | 254 | 62 | ||
More Information
Set Membership: |
|

Lpl
Genomic Map