Assay Details
| Gene Symbol: LOC100773525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507764.1 | XP_003507812.1 | 1 - 2 | 46 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773525| Gene Symbol: LOC100773525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507764.1 | XP_003507812.1 | 1 - 2 | 46 | 77 | |
Set Membership: |
|