Assay Details
| Gene Symbol: Asb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502127.2 | XP_003502175.1 | 7 - 8 | 932 | 71 | |
| XM_007642749.1 | XP_007640939.1 | 9 - 10 | 1526 | 71 | ||
| XM_007642750.1 | XP_007640940.1 | 7 - 8 | 1007 | 71 | ||
| XM_007642751.1 | XP_007640941.1 | 9 - 10 | 1526 | 71 | ||
More Information
Set Membership: |
|

Asb15
Genomic Map