Assay Details
| Gene Symbol: Fam134c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504659.2 | XP_003504707.1 | 3 - 4 | 468 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam134c| Gene Symbol: Fam134c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504659.2 | XP_003504707.1 | 3 - 4 | 468 | 55 | |
Set Membership: |
|