Assay Details
| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007638719.1 | XP_007636909.1 | 4 - 5 | 560 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem38a| Gene Symbol: Tmem38a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007638719.1 | XP_007636909.1 | 4 - 5 | 560 | 67 | |
Set Membership: |
|