Assay Details
| Gene Symbol: Fmo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646019.1 | XP_007644209.1 | 5 - 6 | 768 | 60 | |
| XM_007646020.1 | XP_007644210.1 | 4 - 5 | 628 | 60 | ||
| XM_007646021.1 | XP_007644211.1 | 5 - 6 | 768 | 60 | ||
| GenBank mRNA | JI893756.1 | - | 2 - 3 | 285 | 60 | |
More Information
Set Membership: |
|

Fmo4
Genomic Map