Assay Details
| Gene Symbol: Mettl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502980.2 | XP_003503028.1 | 2 - 3 | 470 | 63 | |
| GenBank mRNA | JI870050.1 | - | 1 - 2 | 343 | 63 | |
| JP039539.1 | - | 1 - 2 | 343 | 63 | ||
More Information
Set Membership: |
|

Mettl2a
Genomic Map