Assay Details
| Gene Symbol: Coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509934.2 | XP_003509982.1 | 6 - 7 | 861 | 59 | |
| XM_007650579.1 | XP_007648769.1 | 7 - 8 | 1003 | 59 | ||
| XM_007650580.1 | XP_007648770.1 | 7 - 8 | 1010 | 59 | ||
| XM_007650581.1 | XP_007648771.1 | 7 - 8 | 1037 | 59 | ||
| XM_007650582.1 | XP_007648772.1 | 6 - 7 | 868 | 59 | ||
| XM_007650584.1 | XP_007648774.1 | 7 - 8 | 994 | 59 | ||
| XM_007650585.1 | XP_007648775.1 | 7 - 8 | 1044 | 59 | ||
| XM_007650586.1 | XP_007648776.1 | 7 - 8 | 987 | 59 | ||
| XM_007650587.1 | XP_007648777.1 | 6 - 7 | 852 | 59 | ||
| XM_007650588.1 | XP_007648778.1 | 6 - 7 | 845 | 59 | ||
| GenBank mRNA | JI887431.1 | - | 7 - 8 | 986 | 59 | |
| JP057927.1 | - | 7 - 8 | 992 | 59 | ||
More Information
Set Membership: |
|

Coro1b
Genomic Map