Assay Details
| Gene Symbol: Srsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007624205.1 | XP_007622395.1 | 1 - 2 | 357 | 60 | |
| GenBank mRNA | AB604328.1 | - | 1 - 2 | 191 | 60 | |
| AB604329.1 | - | 1 - 2 | 191 | 60 | ||
More Information
Set Membership: |
|

Srsf1
Genomic Map