Assay Details
| Gene Symbol: LOC103163961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643168.1 | XP_007641358.1 | 3 - 4 | 1276 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103163961| Gene Symbol: LOC103163961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643168.1 | XP_007641358.1 | 3 - 4 | 1276 | 63 | |
Set Membership: |
|