Assay Details
| Gene Symbol: Arl6ip5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495211.2 | XP_003495259.1 | 2 - 3 | 445 | 64 | |
| GenBank mRNA | JI894131.1 | - | 2 - 3 | 426 | 64 | |
| JP060536.1 | - | 2 - 3 | 426 | 64 | ||
More Information
Set Membership: |
|

Arl6ip5
Genomic Map