Assay Details
| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505949.2 | XP_003505997.2 | 7 - 8 | 1192 | 76 | |
| XM_007646446.1 | XP_007644636.1 | 7 - 8 | 844 | 76 | ||
| GenBank mRNA | JP053608.1 | - | 3 - 4 | 435 | 76 | |
More Information
Set Membership: |
|

Acta2
Genomic Map