Assay Details
| Gene Symbol: Otub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506930.2 | XP_003506978.1 | 3 - 4 | 407 | 69 | |
| XM_007647471.1 | XP_007645661.1 | 3 - 4 | 373 | 69 | ||
| GenBank mRNA | JI895924.1 | - | 2 - 3 | 69 | ||
| JP038448.1 | - | 2 - 3 | 526 | 69 | ||
More Information
Set Membership: |
|

Otub2
Genomic Map