Assay Details
| Gene Symbol: Fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649191.1 | XP_007647381.1 | 2 - 3 | 520 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam155b| Gene Symbol: Fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649191.1 | XP_007647381.1 | 2 - 3 | 520 | 56 | |
Set Membership: |
|