Assay Details
| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496044.2 | XP_003496092.1 | 5 - 6 | 1372 | 60 | |
| GenBank mRNA | JI890155.1 | - | 5 - 6 | 1358 | 60 | |
| JP064333.1 | - | 5 - 6 | 1357 | 60 | ||
More Information
Set Membership: |
|

Tapbpl
Genomic Map