Assay Details
| Gene Symbol: Xpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500090.2 | XP_003500138.1 | 3 - 4 | 593 | 76 | |
| XM_007640824.1 | XP_007639014.1 | 3 - 4 | 597 | 76 | ||
| XM_007640825.1 | XP_007639015.1 | 3 - 4 | 598 | 76 | ||
| XM_007640826.1 | XP_007639016.1 | 3 - 4 | 593 | 76 | ||
| GenBank mRNA | JI880360.1 | - | 2 - 3 | 202 | 76 | |
| JP050693.1 | - | 2 - 3 | 201 | 76 | ||
| S74024.1 | - | 1 - 2 | 44 | 76 | ||
More Information
Set Membership: |
|

Xpa
Genomic Map