Assay Details
| Gene Symbol: Gnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504835.2 | XP_003504883.1 | 7 - 8 | 876 | 67 | |
| XM_007645330.1 | XP_007643520.1 | 7 - 8 | 813 | 67 | ||
More Information
Set Membership: |
|

Gnat3
Genomic Map