Assay Details
| Gene Symbol: Mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509012.2 | XP_003509060.1 | 3 - 4 | 1680 | 72 | |
| XM_007649642.1 | XP_007647832.1 | 4 - 5 | 1037 | 72 | ||
| XM_007649643.1 | XP_007647833.1 | 4 - 5 | 1260 | 72 | ||
| XM_007649644.1 | XP_007647834.1 | 4 - 5 | 1052 | 72 | ||
| XM_007649645.1 | XP_007647835.1 | 4 - 5 | 950 | 72 | ||
| XM_007649646.1 | XP_007647836.1 | 4 - 5 | 1173 | 72 | ||
| XM_007649647.1 | XP_007647837.1 | 4 - 5 | 965 | 72 | ||
More Information
Set Membership: |
|

Mtfp1
Genomic Map