Assay Details
| Gene Symbol: March8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513083.2 | XP_003513131.1 | 6 - 7 | 715 | 64 | |
| XM_007653444.1 | XP_007651634.1 | 7 - 8 | 1577 | 64 | ||
| XM_007653445.1 | XP_007651635.1 | 6 - 7 | 1373 | 64 | ||
| GenBank mRNA | JI869308.1 | - | 2 - 3 | 325 | 64 | |
| JP059867.1 | - | 2 - 3 | 325 | 64 | ||
More Information
Set Membership: |
|

March8
Genomic Map