Assay Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506411.2 | XP_003506459.1 | 1 - 2 | 138 | 69 | |
| GenBank mRNA | JI888046.1 | - | 1 - 2 | 119 | 69 | |
| JP060671.1 | - | 1 - 2 | 119 | 69 | ||
More Information
Set Membership: |
|

Psma2
Genomic Map