Assay Details
| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500104.2 | XP_003500152.1 | 10 - 11 | 2448 | 54 | |
| XM_007640829.1 | XP_007639019.1 | 10 - 11 | 2451 | 54 | ||
| XM_007640830.1 | XP_007639020.1 | 10 - 11 | 2415 | 54 | ||
| XM_007640831.1 | XP_007639021.1 | 9 - 10 | 1949 | 54 | ||
| XM_007640832.1 | XP_007639022.1 | 10 - 11 | 2033 | 54 | ||
| XM_007640833.1 | XP_007639023.1 | 8 - 9 | 2012 | 54 | ||
| GenBank mRNA | JP045429.1 | - | 2 - 3 | 209 | 54 | |
More Information
Set Membership: |
|

Tbc1d2
Genomic Map