Assay Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512969.2 | XP_003513017.2 | 2 - 3 | 160 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam19a3| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512969.2 | XP_003513017.2 | 2 - 3 | 160 | 79 | |
Set Membership: |
|