Assay Details
| Gene Symbol: Gpr39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497614.2 | XP_003497662.1 | 1 - 2 | 1263 | 83 | |
| XM_007614812.1 | XP_007613002.1 | 2 - 3 | 1224 | 83 | ||
More Information
Set Membership: |
|

Gpr39
Genomic Map