Assay Details
| Gene Symbol: Mtss1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501073.2 | XP_003501121.1 | 3 - 4 | 252 | 56 | |
| XM_007641759.1 | XP_007639949.1 | 3 - 4 | 250 | 56 | ||
| XM_007641760.1 | XP_007639950.1 | 3 - 4 | 249 | 56 | ||
| XM_007641761.1 | XP_007639951.1 | 3 - 4 | 250 | 56 | ||
More Information
Set Membership: |
|

Mtss1l
Genomic Map