Assay Details
| Gene Symbol: Tyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510337.2 | XP_003510385.1 | 6 - 7 | 1487 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tyrp1| Gene Symbol: Tyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510337.2 | XP_003510385.1 | 6 - 7 | 1487 | 84 | |
Set Membership: |
|