Assay Details
| Gene Symbol: Hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513676.2 | XP_003513724.2 | 2 - 3 | 218 | 75 | |
| XM_007653927.1 | XP_007652117.1 | 2 - 3 | 217 | 75 | ||
| GenBank mRNA | JI880312.1 | - | 2 - 3 | 205 | 75 | |
| JP037605.1 | - | 2 - 3 | 208 | 75 | ||
More Information
Set Membership: |
|

Hddc3
Genomic Map