Assay Details
| Gene Symbol: Kif4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649715.1 | XP_007647905.1 | 24 - 25 | 3130 | 62 | |
| GenBank mRNA | JI883644.1 | - | 21 - 22 | 2413 | 62 | |
| JP064621.1 | - | 21 - 22 | 2414 | 62 | ||
More Information
Set Membership: |
|

Kif4a
Genomic Map