Assay Details
| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645054.1 | XP_007643244.1 | 1 - 2 | 234 | 78 | |
| GenBank mRNA | JP062221.1 | - | 1 - 2 | 234 | 78 | |
More Information
Set Membership: |
|

Ccdc43
Genomic Map