Assay Details
| Gene Symbol: N6amt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500427.2 | XP_003500475.1 | 3 - 4 | 327 | 102 | |
| XM_007641167.1 | XP_007639357.1 | 3 - 4 | 567 | 102 | ||
| XM_007641168.1 | XP_007639358.1 | 3 - 4 | 451 | 102 | ||
| XM_007641169.1 | XP_007639359.1 | 3 - 4 | 298 | 102 | ||
| XM_007641170.1 | XP_007639360.1 | 3 - 4 | 482 | 102 | ||
| XM_007641171.1 | XP_007639361.1 | 2 - 3 | 192 | 102 | ||
| GenBank mRNA | JI879708.1 | - | 3 - 4 | 279 | 102 | |
More Information
Set Membership: |
|

N6amt2
Genomic Map